Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium hydroxide monohydr25G
Catalog No. 502897779
Encompass
Lithium hydroxide monohydrate is a lithium salt. Crystal structure study reveals it has monoclinic structure with space group C2/m. Its vibrational spectra has been studied using IR Raman and inelastic neutron scattering (INS) spectroscopy. It is reported to transform the surface of titania monoliths to form flower like structures.
Catalog No. 50-289-7779 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 40297425G
Lithium hydroxide monohydrate is a lithium salt. Crystal structure study reveals it has monoclinic structure with space group C2/m. Its vibrational spectra has been studied using IR Raman and inelastic neutron scattering (INS) spectroscopy. It is reported to transform the surface of titania monoliths to form flower like structures.