Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium hydroxide monohydrate

Catalog No. 25427410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-427-410G 10 g
25-427-4250G 250 g
25-427-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-427-410G Supplier Sigma Aldrich Supplier No. 25427410G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Li-OH monohydrate may be used as a precursor for the synthesis of various lithium halides and fatty acids (lithium soaps).Lithium hydroxide monohydrate (Li-OH monohydrate) is prepared industrially by recovering it from spodumene or by evaporating a solution prepared by reacting calcium hydroxide and lithium carbonate. Li-OH monohydrate absorbs atmospheric carbon dioxide, forming lithium carbonate.

Specifications

CAS 1310-66-3
Quantity 10 g
Molecular Formula HLiO -+ H2O
Linear Formula LiOH -+ H2O
MDL Number MFCD00149772
Synonym Lithine hydrate
Molecular Weight (g/mol) 41.96
Percent Purity 99.95% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.