Learn More
Hach Company Lithium Hydroxide Powder Pillows, pk/100
Supplier:  Hach Company 1416369
Encompass
For CO2 absorption in BOD determinations with manometric BOD apparatus, For use with Respirometric BODTrack. Pack of 100 powder pillows.
Catalog No. 50-199-4939