Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium hydroxide reagent500G
Catalog No. 502903117
Encompass
Lithium hydroxide (LiOH) is an alkali metal hydroxide. Lithium chloride SOLUTIONon in water on electrolysis forms LiOH. In respiratory apparatus and submarines it is utilized to uptake carbon dioxide. A study on the redox mechanism of titanium dioxide (TiO2) using cyclic voltammetry X-ray diffraction X-ray photoelectron spectroscopy (XPS) and fourier transform infrared spectroscopy (fTIR) in the aqueous LiOH electrolyte has been reported.
Catalog No. 50-290-3117 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 442410500GA
Lithium hydroxide (LiOH) is an alkali metal hydroxide. Lithium chloride SOLUTIONon in water on electrolysis forms LiOH. In respiratory apparatus and submarines it is utilized to uptake carbon dioxide. A study on the redox mechanism of titanium dioxide (TiO2) using cyclic voltammetry X-ray diffraction X-ray photoelectron spectroscopy (XPS) and fourier transform infrared spectroscopy (fTIR) in the aqueous LiOH electrolyte has been reported.