Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Iodide, MP Biomedicals

Catalog No. ICN15525950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15525950 50 g
ICN15525910 10 g
ICN15525983 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15525950 Supplier MP Biomedicals, Inc Supplier No. 0215525950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Produces a hazy solution at 50% (w/v) in H2O

  • Molecular Formula: LiI
  • Molecular Weight: 133.84
  • Melting Point: 446 °C
  • Boiling Point: 1170 °C
  • Density: 3.49 g/mL
  • Application: Controls regioselectivity in palladium-catalyzed allylic alkylation reactions
  • Safety: H315-H319; Wear dust mask, eyeshields, and gloves
  • Storage: Room temperature
  • Usage: Research or further manufacturing use only
  • Produces a hazy solution at 50% (w/v) in H2O
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium(1+) iodide
SMILES [Li+].[I-]

Specifications

Boiling Point 1170°C
Melting Point 446°C
Quantity 50 g
Linear Formula LiI
Solubility Information Soluble in water
Formula Weight 133.8
Grade Research
Concentration or Composition (by Analyte or Components) 50% (w/v) in H2O
Density 3.49 g/mL
Chemical Name or Material Lithium Iodide
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.