Learn More
Strem, An Ascensus Company CAS# 7790-22-9. 250g. Lithium iodide trihydrate, 99+%. MFCD00149770
SDP
Supplier:  Strem, An Ascensus Company 930381250G
Encompass
CAS# 7790-22-9. 250g. Lithium iodide trihydrate, 99+%. MFCD00149770. Molecular Weight: 133.84 (187.89). Molecular Formula: LiI∙3H2O. Color/form: colorless to yellow xtl. Strem# 93-0381. http://www.strem.com/catalog/v/93-0381/
Catalog No. 50-901-13681