Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA4285118
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4285118 50 g
AA4285104 2 g
AA4285109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4285118 Supplier Thermo Scientific Chemicals Supplier No. 04285118
Add to Cart

missing translation for 'validMainframeMsg'

Light Sensitive and Hygroscopic | CAS: 10377-51-2 | ILi | 133.84 g/mol

Lithium iodide is used as a absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions. Lithium iodide is also a catalyzed conjugate addition reaction of β-dicarbonyl compounds. It is also useful for tetrathioquinodimethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
SMILES [Li+].[I-]

Specifications

Boiling Point 1170°C to 1190°C
Melting Point 449°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water,alcohol,acetone,methanol and ammonium hydoxide.
IUPAC Name lithium;iodide
Formula Weight 133.84
Odor Odorless
Refractive Index 1.955
Sensitivity Hygroscopic; Light sensitive
Density 4.076
Chemical Name or Material Lithium iodide, ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-822-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.