Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Lanthanum Titanate (llto) Powder Battery Grade

Catalog No. 91609950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-609-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-609-950G Supplier Sigma Aldrich Supplier No. 91609950G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Solid electrolytes (SEs) have become a practical option for lithium ion and lithium metal batteries because of their improved safety over commercially available ionic liquids. Solid electrolyte materials are highly conductive and made for advanced solid-state lithium ion battery technologies. These solutions are high purity and battery grade thus making them suitable as standards in LIB research.

Specifications

Molecular Formula LiLaTiO4
Linear Formula LiLaTiO4
Quantity 50 g
Synonym LLTO
Molecular Weight (g/mol) 257.71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.