Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Manganese Nickel Oxide Spinel (lmno) Powder Battery Grade

Catalog No. 91543250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-543-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-543-250G Supplier Sigma Aldrich Supplier No. 91543250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium manganese nickel oxide is a cathode material that can be charged at high voltage. Due to the high potential, the material has a higher energy density compared to lithium cobalt oxide and lithium iron phosphate.NANOMYTE is a registered trademark of NEI Corporation

Specifications

CAS 12031-75-3
Molecular Formula LiMn1.5 Ni0.5O4
Linear Formula LiMn1.5 Ni0.5O4
Quantity 50 g
Synonym LMNO; NANOMYTE™ SP-10
Molecular Weight (g/mol) 182.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.