Learn More
Strem, An Ascensus Company CAS# 13453-69-5. 250g. Lithium metaborate, anhydrous, (99.9%-Li). MFCD00011089
SDP
Supplier:  Strem, An Ascensus Company 930334250G
Encompass
CAS# 13453-69-5. 250g. Lithium metaborate, anhydrous, (99.9%-Li). MFCD00011089. Molecular Weight: 49.75. Molecular Formula: LiBO2. Color/form: white pwdr. Strem# 93-0334. http://www.strem.com/catalog/v/93-0334/
Catalog No. 50-901-13629