Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium metaborate, dried, 99%

Catalog No. AAA1614536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1614536 500 g
AAA1614514 25 g
AAA1614522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1614536 Supplier Thermo Scientific Chemicals Supplier No. A1614536
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol

Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

Solubility
Soluble in water(27.5g/L).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alkali metals, acid anhydrides, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-69-5
Molecular Formula B4H14Li2O7
Molecular Weight (g/mol) 183.23
MDL Number MFCD00011089
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
PubChem CID 123308
IUPAC Name lithium;oxido(oxo)borane
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

Specifications

Melting Point 845°C
Quantity 500 g
Linear Formula LiBO2
Solubility Information Soluble in water(27.5g/L).
Formula Weight 49.75
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 1.397
Chemical Name or Material Lithium metaborate, dried
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 236-631-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.