Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Lithium Metal Scrubbing Solution For Battery Testing Naphthalene 0.1 M In Thf
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 mL
Description
This is a chemical polishing reagent that can smooth the surface of lithium metal and remove native surface impurities. Treatment of lithium metal with this solution has been shown to improve the cycle life and rate performance of lithium metal batteries. Use of this reagent to treat your lithium foil removes the need for scraping to give you a clean lithium surface and may increase the quality and useful testing time for your half-cells.
Specifications
Specifications
| CAS | 91-2-0--3 |
| Molecular Formula | C10H8 |
| Linear Formula | C10H8 |
| Quantity | 10 mL |
| Molecular Weight (g/mol) | 128.17 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction