Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Modified Silica Nanoparticles Non-porous 400 Nm

Catalog No. 9161531G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-615-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-615-31G Supplier Sigma Aldrich Supplier No. 9161531G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Monodisperse nonporous silica nanoparticles with Lithium ion functionalization have been used in electrolytes and nanosalts. We offer highly monodisperse nonporous silica particles with lithium tertbutoxide, with sizes varying from 200 nm to 600 nm.

Specifications

CAS 7631-86-9
Linear Formula SiO2O(CH2)3 SO3-Li+
Quantity 1 g
Synonym Battery electrolytes; Nanosalt; Silica functionalized li-ion particles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.