Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm

Catalog No. p-7048519
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1074114 25 g
AA1074122 100 g
AA1074136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1074114 Supplier Thermo Scientific Chemicals Supplier No. 01074114
Only null left
Add to Cart

CAS: 12031-63-9 | LiNbO3 | 147.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-63-9
Molecular Formula LiNbO3
Molecular Weight (g/mol) 147.84
MDL Number MFCD00011080
InChI Key GQYHUHYESMUTHG-UHFFFAOYSA-N
Synonym Lithium niobate
PubChem CID 23665650
IUPAC Name lithium;oxido(dioxo)niobium
SMILES [Li+].[O-][Nb](=O)=O

Specifications

Melting Point ∼1275°C
Physical Form Powder
Quantity 25 g
Formula Weight 147.85
Percent Purity 99.998%
Grade Puratronic
Density 4.3 g/mL
Assay (metals basis excluding Ta), Ta <50ppm
Chemical Name or Material Lithium niobium oxide
EINECSNumber 234-755-4
RTECSNumber QT9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.