Learn More
Strem, An Ascensus Company CAS# 7790-69-4. 250g. Lithium nitrate, anhydrous, 99%. MFCD00011094
SDP
Catalog No. 5090113638
Encompass
CAS# 7790-69-4. 250g. Lithium nitrate, anhydrous, 99%. MFCD00011094. Molecular Weight: 68.94. Molecular Formula: LiNO3. Color/form: white pwdr. Strem# 93-0339. http://www.strem.com/catalog/v/93-0339/
Catalog No. 50-901-13638 Supplier Strem, An Ascensus Company Supplier No. 930339250G
CAS# 7790-69-4. 250g. Lithium nitrate, anhydrous, 99%. MFCD00011094. Molecular Weight: 68.94. Molecular Formula: LiNO3. Color/form: white pwdr. Strem# 93-0339. http://www.strem.com/catalog/v/93-0339/