Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Oxalate, MP Biomedicals

Catalog No. ICN215314 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215314 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215314 Supplier MP Biomedicals, Inc Supplier No. 0521531450

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30903-87-8
Molecular Formula C2Li2O4
Molecular Weight (g/mol) 101.898
InChI Key YNQRWVCLAIUHHI-UHFFFAOYSA-L
Synonym lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
PubChem CID 68383
IUPAC Name dilithium;oxalate
SMILES [Li+].[Li+].C(=O)(C(=O)[O-])[O-]

Specifications

Quantity 100 g
Formula Weight 101.9
Chemical Name or Material Lithium Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.