Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium perchlorate ACS re500G
Catalog No. 502898790
Encompass
Lithium perchlorate (LiClO4) is a colorless lithium salt. On the crystallization from its aqueous SOLUTIONon it affords lithium perchlorate trihydrate (LiClO4.3H2O). It can be synthesized by reacting lithium chloride with perchloric acid.
Catalog No. 50-289-8790 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 205281500G
Lithium perchlorate (LiClO4) is a colorless lithium salt. On the crystallization from its aqueous SOLUTIONon it affords lithium perchlorate trihydrate (LiClO4.3H2O). It can be synthesized by reacting lithium chloride with perchloric acid.