Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Perchlorate, ACS Reagent, ≥95.0%, Solstice

Catalog No. 60045114 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-114 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-114 Supplier Solstice Supplier No. 205281500G

missing translation for 'validMainframeMsg'

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-9-7791
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M

Specifications

Melting Point 236°C
Color White
pH 6.0 to 9.5 (at 106.4g/L 25°C)
Physical Form Granules
Quantity 500 g
Assay Percent Range ≥95.0%
Linear Formula LiClO4
UN Number UN1481
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
Solubility Information Soluble in water
SMILES [Li+].O.O.O.[O-][Cl](=O)(=O)=O
IUPAC Name sodium;perchlorate;hydrate
PubChem CID 516899
Formula Weight 106.39g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Product Line Solstice
Appearance Granules
Density 2.420g/cu.cm
Chemical Name or Material Lithium Perchlorate
Show More Show Less
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H315-H319-H335
DOTInformation 5.1/PGII
EINECSNumber 232-237-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.