Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium perchlorate, anhydrous, 99%

Catalog No. p-4407584
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4422509 10 g
AA4422518 50 g
AA4422530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4422509 Supplier Thermo Scientific Chemicals Supplier No. 04422509
Only null left
Add to Cart

For Lithium Battery | CAS: 7791-03-9 | ClH6LiO7 | 160.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-9-7791
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
PubChem CID 23665649
IUPAC Name lithium;perchlorate
SMILES [Li+].[O-]Cl(=O)(=O)=O

Specifications

Boiling Point 400°C (decomposition)
Melting Point 236°C
Color White
Physical Form Granules
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in water,alcohol and ethyl acetate.
Formula Weight 106.4
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystals
Density 2.43 g/cm3 at 20°C
Chemical Name or Material Lithium perchlorate, anhydrous
Show More Show Less
Hazard Category H271-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P283-P302+P352-P304+P340-P305+P351+P338-P306+P360-P312-P332+P313-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 232-237-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.