Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium phosphate

Catalog No. 338893100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-889-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-889-3100G Supplier Sigma Aldrich Supplier No. 338893100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium phosphate is majorly used in the form of a conductive thin-film electrolyte for the fabrication of a variety of high performance energy storage devices such as lithium ion batteries and other lithium based rechargeable batteries.

Specifications

CAS 10377-52-3
Quantity 100 g
Molecular Formula Li3O4P
Linear Formula Li3PO4
MDL Number MFCD00016187
Synonym Lithiophosphate (Li3 PO4); Lithium orthophosphate (Li3 PO4); Trilithium monophosphate; Trilithium orthophosphate
Molecular Weight (g/mol) 115.79
Percent Purity 25-27%NaOH basis (titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.