Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium phosphate, Puratronic™, 99.99% (metals basis)

Catalog No. p-7048520
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1074314 25 g
AA1074322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1074314 Supplier Thermo Scientific Chemicals Supplier No. 01074314
Only null left
Add to Cart

CAS: 10377-52-3 | Li3O4P | 115.79 g/mol

Lithium phosphate batteries batteries are now widely used as a staple for cell phones, laptops and some other gadgets. Used in low-expansion porcelain enamel glazes and polymer intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-52-3
Molecular Formula Li3O4P
Molecular Weight (g/mol) 115.79
MDL Number MFCD00016187
InChI Key TWQULNDIKKJZPH-UHFFFAOYSA-K
Synonym lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
PubChem CID 165867
SMILES [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

Specifications

Melting Point 837°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Poor solubility in water. Insoluble in alcohols.
IUPAC Name trilithium(1+) phosphate
Formula Weight 115.79
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Density 2.54
Assay (metals basis)
Chemical Name or Material Lithium phosphate
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 233-823-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.