Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Pyruvate H2O, MP Biomedicals

Catalog No. ICN209254 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209254 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209254 Supplier MP Biomedicals, Inc Supplier No. 0520925401

Chemical Identifiers

CAS 2922-61-4
Molecular Formula C3H3LiO3
Molecular Weight (g/mol) 93.994
InChI Key OFJHGWPRBMPXCX-UHFFFAOYSA-M
Synonym lithium pyruvate, lithium 2-oxopropanoate, pyruvic acid lithium salt, propanoic acid, 2-oxo-, lithium salt, lithiumpyruvat, lithotab 2-oxopropanoate, propanoic acid, 2-oxo-, lithium salt 1:1, c3h3o3.li, lithium pyruvate h2o, lithium 1+ pyruvate
PubChem CID 23663618
IUPAC Name lithium;2-oxopropanoate
SMILES [Li+].CC(=O)C(=O)[O-]

Specifications

Quantity 1 g
Formula Weight 94
Chemical Name or Material Lithium Pyruvate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.