Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Standard, 3000 mEq/L (3 N), Ricca Chemical
SDP

Catalog No. 441016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4410-16 500 mL Natural Poly Bottle
4410-4 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4410-16 Supplier Ricca Chemical Company Supplier No. 441016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-41-8 , 7732-18-5
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 3000 mEq/L,3Normal
CAS Min % 87.0, 11.28
CAS Max % 90.0, 11.74
Physical Form Liquid
Color Colorless
pH Approximately 7
Melting Point Approximately 0°C
Quantity 500 mL
Chemical Name or Material Lithium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.