Learn More
Lithium Standard, For IC, 1,000 mg/L Li+ in Nitric Acid, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Chemical Name or Material | Lithium |
| CAS | 554-13-2, 7697-37-2 |
| Product Line | TraceCERT |
| Concentration | 1000 mg/L Li + in nitric acid |
| Technique | Ion Chromatography |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Nitric Acid |
| Industry Type | Environmental Testing |
| Quantity | 100 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.