Learn More
Medchemexpress LLC Lithium stearate 25g
SDP
Catalog No. 5000206804
Encompass_Preferred
Lithium stearate (Stearic acid (lithium)) is a compound that forms vesicles in reaction with silver nitrate thereby changing the reaction kinetics of the nucleation and self-assembly process of silver stearate It has the activity that even in the presence of excess silver nitrate only about 80% can be converted into silver stearate and the residual lithium stearate will inhibit the growth process of silver stearate crystals thereby producing silver stearate crystals that are much smaller than those obtained from sodium stearate providing an opportunity to further control the self-assembly and crystal growth of silver stearate
Catalog No. 50-002-06804 Supplier Medchemexpress LLC Supplier No. 25GHYW099571
Lithium stearate (Stearic acid (lithium)) is a compound that forms vesicles in reaction with silver nitrate thereby changing the reaction kinetics of the nucleation and self-assembly process of silver stearate It has the activity that even in the presence of excess silver nitrate only about 80% can be converted into silver stearate and the residual lithium stearate will inhibit the growth process of silver stearate crystals thereby producing silver stearate crystals that are much smaller than those obtained from sodium stearate providing an opportunity to further control the self-assembly and crystal growth of silver stearate