Learn More
Medchemexpress LLC Lithium sulfate 50g
SDP
Catalog No. 5000205851
Encompass_Preferred
Lithium sulfate is an inorganic compound that can improve the cycling life and efficiency of a 2 V/20 A H lead-acid battery Lithium sulfate can be used to study mental illnesses[1][2]
Catalog No. 50-002-05851 Supplier Medchemexpress LLC Supplier No. HYW09446450G
Lithium sulfate is an inorganic compound that can improve the cycling life and efficiency of a 2 V/20 A H lead-acid battery Lithium sulfate can be used to study mental illnesses[1][2]