Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate, Anhydrous, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18611338 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-338 25 g Poly Bottle
18-611-343 2.5 kg Poly Bottle
18-611-340 12 kg Poly Pail
18-611-342 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-338 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L112425GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
  • Chemical Name: Lithium sulfate, anhydrous
  • Formula: Li2SO4
  • Molecular Weight: 109.94 g/mole
  • Assay (Current ACS Titration Method): 98.5%
  • Physical State: Solid
  • Solubility: Soluble in cold water
  • Stability: Hygroscopic; keep container tightly closed. Stable under recommended storage conditions.
  • Reactivity: Reactive with oxidizing agents; loses water above 120°C.
  • Melting Point: 845°C to 859°C
  • Specific Gravity: 2.22

Chemical Identifiers

CAS 10377-48-7

Specifications

Melting Point 845°C to 859°C
Color White
CAS Max % 1
Physical Form Solid
Quantity 25 g
Assay Percent Range 98.50%
Molecular Formula Li2SO4
UN Number NONH for all modes of transport
Synonym Lithium Sulphate, Sulfuric Acid, Dilithium SaltSulfuric Acid, Lithium Salt (1:2)
Solubility Information Soluble in cold water
SMILES [Li+].[Li+].[O-]S(=O)(=O)[O-]
IUPAC Name Dilithium sulfate
Molecular Weight (g/mol) 109.94
Formula Weight 109.94 g/mol
Percent Purity 98.5%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Poly Bottle
Sensitivity Hygroscopic
Specific Gravity 2.22
Decomposition Information Sulfur Oxides, Lithium Oxides
Chemical Name or Material Lithium Sulfate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.