Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate, Anhydrous, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18611343 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-343 2.5 kg Poly Bottle
18-611-338 25 g Poly Bottle
18-611-340 12 kg Poly Bottle
18-611-342 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-343 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L11242.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity anhydrous reagent grade lithium sulfate for use in chemical research and analysis.

  • Chemical Name: Lithium sulfate, anhydrous
  • Formula: Li2SO4
  • Assay (Current ACS Titration Method): 98.5%
  • Physical State: Solid
  • Stability: Hygroscopic; keep container tightly closed. Stable under recommended storage conditions.
  • Melting Point: 845°C to 859°C
  • Specific Gravity: 2.22
  • CAS Number: 10377-48-7
  • Appearance: White, odorless solid
  • Reactivity: Reactive with oxidizing agents; loses all water at temperatures above 120°C
  • Chemical Formula: Li2SO4
  • Physical Form: Crystalline Powder
  • Chemical Stability: Stable under recommended storage conditions.
  • Incompatible Materials: Oxidizing agents, moisture.
  • Grade: Reagent
  • Purity: 98.5%
  • Color: White
  • Odor: Odorless
  • Solubility: Soluble in cold water
  • Molecular Weight: 109.94 g/mole

Chemical Identifiers

CAS 10377-48-7

Specifications

Melting Point 845°C to 859°C
Color White
CAS Max % 1
Physical Form Adhering Powder and/or Lumps
Quantity 2.5 kg
Assay Percent Range 98.50%
Molecular Formula Li2SO4
UN Number NONH for all modes of transport
Synonym Lithium Sulphate, Sulfuric Acid, Dilithium SaltSulfuric Acid, Lithium Salt (1:2)
Solubility Information Soluble in cold water
SMILES [Li+].[Li+].[O-]S(=O)(=O)[O-]
IUPAC Name Dilithium sulfate
Molecular Weight (g/mol) 109.94
Formula Weight 109.94 g/mol
Percent Purity 98.5%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Poly Bottle
Sensitivity Hygroscopic
Specific Gravity 2.22
Chemical Name or Material Lithium Sulfate, Anhydrous
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.