Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate Monohydrate, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15253990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15253990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15253990 Supplier MP Biomedicals, Inc Supplier No. 0215253990
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent grade chemical suitable for analytical and laboratory applications.

  • Grade: ACS reagent
  • Molecular Formula: H2Li2O5S
  • Molecular Weight: 127.96 g/mol
  • CAS Number: 10102-25-7
  • Melting Point: 130 °C
  • Density: 2.1 g/mL
  • Solubility: Soluble in cold water
  • Storage: Room temperature
  • Hazard Statement: H302
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-25-7
Molecular Formula H2Li2O5S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00149766
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
PubChem CID 165815
IUPAC Name dilithium(1+) hydrate sulfate
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Li2SO4·H2O
Solubility Information Soluble in water
ChEBI CHEBI:63300
Formula Weight 128
Grade Meets ACS reagent chemical requirements
Sensitivity Hygroscopic
Chemical Name or Material Lithium Sulfate Monohydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.