Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate, Monohydrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611344 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-344 25 g Poly Bottle
18-611-345 125 g Poly Bottle
18-611-346 500 g Poly Bottle
18-611-347 2.5 kg Poly Pail
18-611-348 12 kg Poly Pail
18-611-349 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-344 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L112525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
  • Grade: ACS
  • Chemical Formula: Li2SO4·H2O
  • Molecular Weight: 127.96 g/mol
  • Appearance: Crystalline powder
  • Melting Point: 880 °C
  • Solubility: Soluble in cold water; Insoluble in acetone
  • Stability: Hygroscopic; stable under recommended storage conditions
  • Hazard Classification: Harmful if swallowed

Chemical Identifiers

CAS 10102-25-7

Specifications

Melting Point 880°C
Color White
CAS Max % 1
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Molecular Formula Li2SO4·H2O
UN Number NONH for all modes of transport
Synonym Lithium Mesosulfate, Sulfuric Acid, Dilithium Salt, Monohydrate
Solubility Information Soluble in cold water, Insoluble in Acetone
InChI Formula InChI=1S/Li.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+1;;/p-2
SMILES [Li+].[Li+].[O-]S(=O)(=O)[O-].O
IUPAC Name Dilithium sulfate monohydrate
Molecular Weight (g/mol) 127.96
Formula Weight 127.96
Percent Purity 99%
Grade ACS
Concentration or Composition (by Analyte or Components) Lithium Sulfate, Monohydrate
Manufacturing Origin USA
Identification Passes Test
Loss on Drying 13.0-15.0%
Packaging Poly Bottle
Sensitivity Hygroscopic
Appearance Crystalline Powder
Specific Gravity 2.06
Decomposition Information Sulfur Oxides, Lithium Oxides
Chemical Name or Material Lithium Sulfate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.