Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate, Monohydrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611349 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-349 45 kg Poly Bottle
18-611-344 25 g Poly Bottle
18-611-345 125 g Poly Bottle
18-611-346 500 g Poly Bottle
18-611-347 2.5 kg Poly Bottle
18-611-348 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-349 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L112545KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High-purity ACS grade lithium sulfate monohydrate suitable for laboratory and analytical applications.

  • Chemical Formula: Li2SO4·H2O
  • Molecular Weight: 127.96 g/mol
  • Appearance: Crystalline powder
  • Stability: Hygroscopic; stable under recommended storage conditions
  • Formula: Li2SO4·H2O
  • Loss on Drying: 13.0-15.0%
  • Flammability: Non-flammable
  • Purity: 99%
  • Physical State: Solid
  • Color: White
  • Solubility: Soluble in cold water; Insoluble in Acetone
  • Assay: 99%
  • Form: Crystal
  • CAS number: 10102-25-7
  • Grade: ACS
  • Melting Point: 880 °C
  • Specific Gravity: 2.06
  • Hazard Classification: Acute toxicity - Oral - Category 4; Harmful if swallowed

Chemical Identifiers

CAS 10102-25-7

Specifications

Melting Point 880°C
Color White
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Assay Percent Range 99%
Molecular Formula Li2SO4·H2O
Solubility Information Soluble in cold water, Insoluble in acetone
SMILES [Li+].[Li+].[O-]S(=O)(=O)[O-].O
IUPAC Name Dilithium sulfate monohydrate
Molecular Weight (g/mol) 127.96
Formula Weight 127.96
Percent Purity 99%
Grade ACS/Reagent
Manufacturing Origin USA
Loss on Drying 13.0 to 15.0%
Packaging Poly Bottle
Sensitivity Hygroscopic
Appearance Crystalline Powder
Specific Gravity 2.06
Decomposition Information Sulfur Oxides, Lithium Oxides
Chemical Name or Material Lithium Sulfate, Monohydrate, Crystal
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.