Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Sulfate Monohydrate, puriss p.a, ACS, ≥99.0% (dried basis, T), Solstice

Catalog No. 60047230 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-230 100 g Poly bottle
60-047-231 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-230 Supplier Solstice Supplier No. 62609100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.0% (dried basis, T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-25-7
Molecular Formula H2Li2O5S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00149766
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
PubChem CID 165815
IUPAC Name dilithium;sulfate;hydrate
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O

Specifications

Melting Point 236°C
Color White
pH 6.0 to 9.5 (at 104g/L 25°C)
Physical Form Granules
Quantity 100 g
Assay Percent Range ≥99.0%
Linear Formula Li2SO4 · H2O
UN Number NONH for all modes of transport
Solubility Information Soluble in water
Formula Weight 127.96g/mol
Percent Purity ≥99.0% (dried basis, T)
Loss on Drying 13.0 to15.0%
Packaging Poly bottle
Product Line Solstice
Appearance White Powder or Crystals
Assay Dried Basis, T
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less
RTECSNumber 233-820-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.