Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Tert-Butoxide

Catalog No. 400173100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-017-3100G 100 g
40-017-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 40-017-3100G Supplier Sigma Aldrich Supplier No. 400173100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium tert-butoxide (LiOtBu) is a weakly basic and nucleophilic alkali metal oxide commonly used as an initiator for anionic polymerization.Other synthetic applications: In combination with potassium diisopropylamide, LiOtBu can be used to deprotonate 1-(phenylseleno) alkenes and bis (phenylseleno) acetals.LiOtBu can mediate the +a-alkylation reaction of ketones with primary alcohols in the absence of any transition metal catalyst.LiOtBu is an effective base for the synthesis of 3,4,5-trisubstituted 3H-oxazol-2-ones and 3,4-disubstituted (Z)-oxazolidin-2-ones from substituted propargyl alcohols and aryl/alkyl isocyanates using DMF as a solvent.

Specifications

CAS 1907-33-1
Molecular Formula C4H9LiO
Linear Formula (CH3)3COLi
MDL Number MFCD00050479
Quantity 100 g
Synonym LiOtBu
Molecular Weight (g/mol) 80.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.