Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA4413318
Click to view available options
Quantity:
2 g
10 g
50 g
1907-33-1
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
1907-33-1
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]

Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼150°C (decomposition)
Color White to Cream
Odor Odorless
Linear Formula (CH3)3COLi
Quantity 50 g
UN Number UN3206
Beilstein 3620018
Sensitivity Air and moisture sensitive
Solubility Information Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether.
Formula Weight 80.06
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium tert-butoxide
Show More Show Less
Hazard Category H228-H302-H314
Hazard Statement GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 217-611-5
RTECSNumber UB8520000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit