Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Tert-Butoxide Solution

Catalog No. 39819550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-819-550ML 50 mL
39-819-5250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-819-550ML Supplier Sigma Aldrich Supplier No. 39819550ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium tert-butoxide solution is generally used as a strong base in organic synthesis. It can be used:For the synthesis of lithium modified silica nano-particles for conductive gel electrolytes.As a catalyst for ring-opening polymerization of lactides.As a lithium precursor for the synthesis of LiV3O8 nanoparticles by flame spray pyrolysis.

Specifications

CAS 1907-33-1
Density 0.888 g/mL at 25°C
Boiling Point 67°C
Molecular Formula C4H9LiO
Linear Formula (CH3)3COLi
Quantity 50 mL
MDL Number MFCD00050479
Molecular Weight (g/mol) 80.05 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.