Learn More
Malvern Panalytical Li Tetraborate UP, Ultra High-purity (99,995+%), 1 kg bottle
Supplier:  Malvern Panalytical C060100
Encompass
Lithium Tetraborate, High Density, Grade Ultra Pure, 1 KgUltra High-purity (99,995+%) pre-fused flux to prepare samples for XRF or ICP analysis. Composition : 100 % Lithium Tetraborate. 1 kg bottle. Properties: spherical vitreous beads, anhydrous and non-hygroscopic, homogeneous particles, no dust, controlled granulometry, high fluidity.
Catalog No. NC1864471