Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Tetrafluoroborate, MP Biomedicals

Catalog No. ICN221088 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN221088 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN221088 Supplier MP Biomedicals, Inc Supplier No. 0522108810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Chemical Name: Lithium tetrafluoroborate
  • Molecular Formula: BF4Li
  • Formula Weight: 93.74
  • CAS Number: 14283-07-9
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium(1+) tetrafluoroboranuide
SMILES [Li+].F[B-](F)(F)F

Specifications

Quantity 10 g
Linear Formula LiBF4
InChI Formula InChI=1S/BF4.Li/c2-1(3,4)5;/q-1;+1
Formula Weight 93.75
Sensitivity Hygroscopic
Chemical Name or Material Lithium Tetra Fluoroborate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.