Learn More
AdipoGen Lithium tetrakis PFPBEt ethera
SDP
Supplier:  AdipoGen CDXL0011G010
Encompass_Preferred
Chemical. CAS 371162-53-7. Formula C24BF20Li . 2.5 C4H10O. MW 871.28. Synthetic. Salt for the generation of cationic transition metal complexes.
Catalog No. 50-256-0616