Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium titanate, spinel

Catalog No. 70227725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-227-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-227-725G Supplier Sigma Aldrich Supplier No. 70227725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Lithium titanate, spinel nanopowder can be used as an anode material, which shows an ion conductivity of 10-13 Scm-1 at room temperature. It can also be used as an alternative to conventional graphite materials. It can further be used in the fabrication of high-performance lithium-ion batteries for electric vehicles (EVs).Product of Engi-Mat Co.

Specifications

CAS 12031-95-7
Molecular Formula Li4O12Ti5
Linear Formula Li4Ti5O12
Quantity 25 g
MDL Number MFCD11656084
Synonym LTO nanopowder; Lithium titanate spinel oxide; Lithium titanium oxide
Molecular Weight (g/mol) 459.09
Percent Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.