Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. aa43324ab
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43324AB 10 mL
AA43324AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43324AB Supplier Thermo Scientific Chemicals Supplier No. 043324AB
Only null left
Add to Cart

CAS: 17476-04-9 | C12H28AlLiO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17476-04-9
Molecular Formula C12H28AlLiO3
MDL Number MFCD00011532
Synonym Lithium hydridotris(tert-butoxy)aluminate

Specifications

Color Colorless
Density 0.904 g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula Li[(CH3)3CO]3AlH
Quantity 10 mL
UN Number UN3399
Sensitivity Moisture sensitive
Solubility Information It is soluble in organic solvents at low temperatures
Formula Weight 254.28
Physical Form Liquid
Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride
Show More Show Less
Hazard Category H225-H260-H302-H315-H318-H335-H336-H351
Hazard Statement GHS H Statement
H225-H260-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.