Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Tri-Tert-Butoxyaluminum Hydride Solution

Catalog No. 241814100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-181-4100ML 100 mL
24-181-4800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-181-4100ML Supplier Sigma Aldrich Supplier No. 241814100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium tri-tert-butoxyaluminum hydride is a mild reducing agent mainly used for the selective reduction of ketones.Reducing agent

Specifications

CAS 17476-04-9
Density 0.904 g/mL at 25°C
Molecular Formula C12H28AlLiO3
Linear Formula LiAlH[OC(CH3)3]3
Quantity 100 mL
MDL Number MFCD00011532
Molecular Weight (g/mol) 254.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.