Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium triethylborohydride, 1M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Specifications
| Color | Colorless |
| Density | 0.892 g/mL |
| Flash Point | −17°C (1°F) |
| Packaging | ChemSeal™ bottles |
| Linear Formula | LiB(CH2CH3)3H |
| Quantity | 500 mL |
| UN Number | UN3399 |
| Beilstein | 4151240 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,5544 |
| Show More |
Safety and Handling
| Hazard Category | H225-H250-H260-H302-H314-H335-H336-H351 |
| Hazard Statement | GHS H Statement H225-H260-H314-H318 Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
| TSCA | No |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?