Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium (Trimethylsilyl)Acetylide Solution

Catalog No. 33545225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-545-225ML 4 x 25 mL
33-545-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 33-545-225ML Supplier Sigma Aldrich Supplier No. 3354524X25ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium (trimethylsilyl)acetylide can be used as a reagent: In the transmetalation and nucleophilic displacement reactions. To prepare propargylic alcohol derivatives by reacting with aldehydes and ketones. To synthesize +a,+aynones by treating with Weinreb amide or with isoxazolidide. Along with Grignard reagent to convert +a- and +a-thiolactams to thioiminium salts, which are employed as key intermediates to produce disubstituted pyrrolidines.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 54655-07-1
Density 0.88 g/mL at 25°C
Molecular Formula C5H9LiSi
Linear Formula (CH3)3SiC///CLi
MDL Number MFCD00075059
Quantity 4 x 25 mL
Synonym (Trimethylsilyl)ethynyllithium
Molecular Weight (g/mol) 104.15 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.