Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium wire, 3.2mm (0.125in) dia, 99.8%, Na 0.03% max

Catalog No. aa1077114
Change view
Click to view available options
Quantity:
25 g
4 x 25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1077114 25 g
AA1077149 4 x 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1077114 Supplier Thermo Scientific Chemicals Supplier No. 01077114
Only null left
Add to Cart

CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]

Specifications

Density 0.53
Quantity 25 g
Form Annealed Wire
Assay Percent Range 99.8%
Formula Weight 6.941
Chemical Name or Material Lithium, 3.2mm (0.125 in.) Dia.
Hazard Category H260-H314
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
EINECSNumber 231-102-5
RTECSNumber OJ5540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.