Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lithocholic acid (3α-Hydroxy-5β-cholanic acid) | 434-13-9 | 100.0% | 25 MG
SDP

Catalog No. 5000451027
Encompass_Preferred

Lithocholic acid is a toxic secondary bile acid that can promote intrahepatic cholestasis and tumorigenesis. It is also an FXR antagonist and a PXR/SXR agonist.

  • Synonym: 3α-Hydroxy-5β-cholanic acid
  • Purity of 99.96%
  • Appears as a solid
  • White to off-white color
  • Classified as a steroid
  • Sourced as an endogenous metabolite and from human gut microbiota
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Exhibits FXR antagonism
  • Inhibits BSEP expression
  • Inhibits neuroblastoma cell proliferation

Catalog No. 50-004-51027 Supplier Medchemexpress LLC Supplier No. 25MGHYB0172R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lithocholic acid is a toxic secondary bile acid that can promote intrahepatic cholestasis and tumorigenesis. It is also an FXR antagonist and a PXR/SXR agonist.

  • Synonym: 3α-Hydroxy-5β-cholanic acid
  • Purity of 99.96%
  • Appears as a solid
  • White to off-white color
  • Classified as a steroid
  • Sourced as an endogenous metabolite and from human gut microbiota
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Exhibits FXR antagonism
  • Inhibits BSEP expression
  • Inhibits neuroblastoma cell proliferation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.