Learn More
Cayman Chemical Lithocholic Acid 3-sulfate (sodium salt), Purity: ≥98%, CAS: 64936-81-8, 10 mg
SDP
Catalog No. 502984818
Encompass_Preferred
A metabolite of lithocholic acid; water soluble and the primary form of lithocholic acid in the bile duct; forms a complex with calcium in vitro; biliary secretion increased in a rat model of streptozotocin-induced diabetes
Catalog No. 50-298-4818 Supplier Cayman Chemical Supplier No. 2067610 MG
A metabolite of lithocholic acid; water soluble and the primary form of lithocholic acid in the bile duct; forms a complex with calcium in vitro; biliary secretion increased in a rat model of streptozotocin-induced diabetes