Learn More
Cayman Chemical LIthocholIc AcId 3sulfatS 50mg
SDP
Catalog No. 502984737
Encompass_Preferred
A metabolite of lithocholic acid; water soluble and the primary form of lithocholic acid in the bile duct; forms a complex with calcium in vitro; biliary secretion increased in a rat model of streptozotocin-induced diabetes
Catalog No. 50-298-4737 Supplier Cayman Chemical Supplier No. 2067650 MG
A metabolite of lithocholic acid; water soluble and the primary form of lithocholic acid in the bile duct; forms a complex with calcium in vitro; biliary secretion increased in a rat model of streptozotocin-induced diabetes