Learn More
Selleck Chemical LLC Lithocholic acid 50mg 434-13-9
SDP
Supplier:  Selleck Chemical LLC S400350MG
Encompass_Preferred
Lithocholic acid is a toxic secondary bile acid, causes intrahepatic cholestasis, has tumor-promoting activity, its toxic effect can be protected after it activates the vitamin D receptor, PXR and FXR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3519