Learn More
Cayman Chemical Lithocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 83701-16-0, 10 mg
SDP
Catalog No. 502980746
Encompass_Preferred
An internal standard for the quantification of lithocholic acid by GC- or LC-MS
Catalog No. 50-298-0746 Supplier Cayman Chemical Supplier No. 2083110 MG
An internal standard for the quantification of lithocholic acid by GC- or LC-MS