Learn More
Cayman Chemical LIthocholIc AcId-d4 500ug
SDP
Catalog No. 502983147
Encompass_Preferred
An internal standard for the quantification of lithocholic acid by GC- or LC-MS
Catalog No. 50-298-3147 Supplier Cayman Chemical Supplier No. 20831500 UG
An internal standard for the quantification of lithocholic acid by GC- or LC-MS